Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Nociceptin

Catalog No. 4879601MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-796-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-796-01MG Supplier MilliporeSigma™ Supplier No. 4879601MG
Add to Cart
Add to Cart

Endogenous ligand for the opioid-like G-protein coupled receptor, ORL1 (Opioid Receptor-Like 1).

Peptide Sequence: H-Phe-Gly-Gly-Phe-Thr-Gly-Ala-Arg-Lys-Ser-Ala-Arg-Lys-Leu-Ala-Asn-Gln-OH
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 98% by HPLC
Target Proteins Opioid-like G-protein coupled receptor, ORL1 (Opioid Receptor-Like 1)
CAS 170713-75-4
Molecular Formula C79H129N27O22
Formula Weight 1809.1g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.