CAS RN 17075-03-5
IUPAC Name:
pyren-4-amine
Synonyms:
pyren-4-amine
Molecular Weight (g/mol):
217.27
Molecular Formula:
C16H11N
InChi Key:
AYBSKADMMKVOIV-UHFFFAOYSA-N
SMILES:
NC1=CC2=CC=CC3=CC=C4C=CC=C1C4=C23
1
–
1
of
1
results
1
4-Aminopyrene 97.0+%, TCI America™
CAS: 17075-03-5 Molecular Formula: C16H11N Molecular Weight (g/mol): 217.27 MDL Number: MFCD00021660 InChI Key: AYBSKADMMKVOIV-UHFFFAOYSA-N Synonym: 4-Pyrenamine PubChem CID: 119443 IUPAC Name: pyren-4-amine SMILES: NC1=CC2=CC=CC3=CC=C4C=CC=C1C4=C23