Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminopyrene 97.0+%, TCI America™
SDP

Catalog No. A21801G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21801G Supplier TCI America Supplier No. A21801G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17075-03-5
Molecular Formula C16H11N
Molecular Weight (g/mol) 217.27
MDL Number MFCD00021660
InChI Key AYBSKADMMKVOIV-UHFFFAOYSA-N
Synonym 4-Pyrenamine
PubChem CID 119443
IUPAC Name pyren-4-amine
SMILES NC1=CC2=CC=CC3=CC=C4C=CC=C1C4=C23

Specifications

Quantity 1 g
Formula Weight 217.27
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminopyrene
RTECSNumber UR2277000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.