accessibility menu, dialog, popup

UserName

Viewing profile as user:
MgBrClF

CAS RN 170793-00-7

IUPAC Name: bromo(4-chloro-3-fluorophenyl)magnesium
Synonyms: bromo(4-chloro-3-fluorophenyl)magnesium
Molecular Weight (g/mol): 233.75
Molecular Formula: C6H3BrClFMg
InChi Key: JVKNCECRGPRLKQ-UHFFFAOYSA-M
SMILES: FC1=C(Cl)C=CC([Mg]Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results