Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Diethyl-1,1,2,3,3,3-Hexafluoropropylamine

Catalog No. 56499025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-499-025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 56-499-025G Supplier Sigma Aldrich Supplier No. 56499025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

GC analysis includes Et2NCF=CFCF3 (two isomers), both of which are active fluorinating agents.Reactant for: Friedel-Crafts type arylation of hydrofluorocarbonsMicrobial hydroxylation of antibioticsElectrochemical fluorination of trialkylamines and tetraalkylammonium saltsFluorination of pyrethroidesReactant for synthesis of: +a-Hydroxy-+a-fluoro-+a-trifluoromethylcarboxamideFlorfenicol and thiamphenicol

Specifications

CAS 170793-00-7
Density 0.968 g/mL at 25°C
Boiling Point 65°C
Molecular Formula C6H3BrClFMg
Refractive Index n20/D 1.3460
Linear Formula ClC6H3(F)MgBr
Quantity 25 g
MDL Number MFCD02260213
Synonym Ishikawa's Reagent
Molecular Weight (g/mol) 233.75 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.