accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1708-41-4

IUPAC Name: 2-(furan-2-yl)-1,3-dioxolane
Synonyms: 2-(furan-2-yl)-1,3-dioxolane
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: FCACHSUJEMZCMK-UHFFFAOYSA-N
SMILES: C1COC(O1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results