Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1,3-Dioxolan-2-yl)furan 98.0+%, TCI America™
SDP

Catalog No. D241725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D241725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D241725G Supplier TCI America Supplier No. D241725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1708-41-4
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.14
MDL Number MFCD00671520
InChI Key FCACHSUJEMZCMK-UHFFFAOYSA-N
Synonym Furfural Ethylene Acetal, 2-(2-Furyl)-1,3-dioxolane
PubChem CID 263158
IUPAC Name 2-(furan-2-yl)-1,3-dioxolane
SMILES C1COC(O1)C1=CC=CO1

Specifications

Color Yellow
Boiling Point 76°C
Quantity 25 g
Formula Weight 140.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(1,3-Dioxolan-2-yl)furan
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.