accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3COSiCH3OSiCH3CH3OSiCH3H3C

CAS RN 17082-46-1

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 265.58
Molecular Formula: C7H21O3Si4
InChi Key: BGISVIDNIBCUTN-UHFFFAOYSA-N
SMILES: C[Si](C)O[Si](C)(O[Si](C)C)O[Si](C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results