Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[(Dimethylsilyl)oxy]-1,1,3,5,5-pentamethyltrisiloxane 96.0+%, TCI America™
SDP

Catalog No. D52635ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52635ML 5 mL
D526325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52635ML Supplier TCI America Supplier No. D52635ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17082-46-1
Molecular Formula C7H21O3Si4
Molecular Weight (g/mol) 265.58
MDL Number MFCD00054789
InChI Key BGISVIDNIBCUTN-UHFFFAOYSA-N
Synonym Methyltris(dimethylsiloxy)silane
PubChem CID 11032875
IUPAC Name [bis[(dimethyl-$l^{3}-silanyl)oxy]-methylsilyl]oxy-dimethylsilicon
SMILES C[Si](C)O[Si](C)(O[Si](C)C)O[Si](C)C

Specifications

Color Colorless
Boiling Point 164°C
Quantity 5 mL
Formula Weight 268.61
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-[(Dimethylsilyl)oxy]-1,1,3,5,5-pentamethyltrisiloxane
EINECSNumber (7)-0477&(7)-0476
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.