accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 170853-04-0

CH3H3CCH3OONHO

CAS RN 170853-04-0

IUPAC Name: tert-butyl N-[(3-formylphenyl)methyl]carbamate
Synonyms: tert-butyl N-[(3-formylphenyl)methyl]carbamate
Molecular Weight (g/mol): 235.28
Molecular Formula: C13H17NO3
InChi Key: JORXNWZTJLYRQA-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=CC=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results