Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl N-(3-formylbenzyl)carbamate, 90%, Thermo Scientific™

Catalog No. MO07494DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO07494DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07494DA Supplier Thermo Scientific Chemicals Supplier No. MO07494DA
Add to Cart

Chemical Identifiers

CAS 170853-04-0
Molecular Formula C13H17NO3
Molecular Weight (g/mol) 235.283
InChI Key JORXNWZTJLYRQA-UHFFFAOYSA-N
PubChem CID 2794834
IUPAC Name tert-butyl N-[(3-formylphenyl)methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC=CC(=C1)C=O

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym tert-butyl 3-formylbenzylcarbamate, tert-butyl n-3-formylphenyl methyl carbamate, tert-butyl 3-formylbenzyl carbamate, 3-aminomethyl benzaldehyde, n-boc protected, 3-formylbenzylamine, n-boc protected, tert-butyl n-3 formylbenzyl carbamate, tert-butyl n-3-formylbenzyl carbamate, tert-butyl3-formylbenzylcarbamate, 3-formylbenzyl carbamic acid tert-butyl ester, 3-formyl-benzyl-carbamic acid tert-butyl ester
Formula Weight 235.28
Percent Purity 90%
Chemical Name or Material tert-Butyl 3-formylbenzylcarbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.