accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SOONNO

CAS RN 170858-33-0

IUPAC Name: 4-{4-[2-(3,4-dihydro-1H-2-benzopyran-1-yl)ethyl]piperazin-1-yl}benzene-1-sulfonamide
Synonyms: 4-{4-[2-(3,4-dihydro-1H-2-benzopyran-1-yl)ethyl]piperazin-1-yl}benzenesulfonamide
Molecular Weight (g/mol): 401.53
Molecular Formula: C21H27N3O3S
InChi Key: WNUQCGWXPNGORO-UHFFFAOYNA-N
SMILES: NS(=O)(=O)C1=CC=C(C=C1)N1CCN(CCC2OCCC3=CC=CC=C23)CC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results