accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17086-28-1

IUPAC Name: (2Z,4S,4aR,5S,5aR,6R,12aS)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-5,10,11,12a-tetrahydroxy-6-methyl-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione hydrate
Synonyms: doxytetracycline hydrate
Molecular Weight (g/mol): 462.46
Molecular Formula: C22H26N2O9
InChi Key: ADAURKPGCFUREH-UQVCFKGQSA-N
SMILES: O.C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H](N(C)C)C(=O)\C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results