CAS RN 17110-51-9
IUPAC Name:
9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl (9Z)-octadec-9-en-1-yl carbonate
Synonyms:
cholesteryl oleyl carbonate
Molecular Weight (g/mol):
681.14
Molecular Formula:
C46H80O3
InChi Key:
XMPIMLRYNVGZIA-PFONDFGANA-N
SMILES:
CCCCCCCC\C=C/CCCCCCCCOC(=O)OC1CCC2(C)C3CCC4(C)C(CCC4C3CC=C2C1)C(C)CCCC(C)C
1
–
1
of
1
results
1
Sigma Aldrich Cholesteryl oleyl carbonate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.