Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cholesteryl oleyl carbonate

Catalog No. 15115725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-115-725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-115-725G Supplier Sigma Aldrich Supplier No. 15115725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cholesteryl oleyl carbonate(COC) is a carbonate ester of cholesterol and oleyl alcohol with carbonic acid that is used as a liquid crystal(LC) molecule. It has is a crystalline material with a helical structure.COC can be incorporated in the polyethylene glycol(PEG)-polyurethane(PU) matrix that exhibits lower thrombogenicity and an improved haemocompatibility which can used as a biomaterial in biomedical applications. It may also be self-assembled with cadmium selenide(CdSe) quantum dots(QDs) for the fabrication of novel multifunctional switchable devices.

Specifications

CAS 17110-51-9
Molecular Formula C46H80O3
Linear Formula C46H80O3
MDL Number MFCD00003634
Quantity 25 g
Synonym 3 beta-Hydroxy-5-cholestene 3-oleyl carbonate; 5-Cholesten-3 beta-ol 3-oleyl carbonate; 5-Cholesten-3-yl 9-octadecenyl carbonate; Cholesterol 9-octadecenyl carbonate
Molecular Weight (g/mol) 681.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.