accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2Cl

CAS RN 1712-70-5

IUPAC Name: 1-chloro-4-(prop-1-en-2-yl)benzene
Synonyms: 1-chloro-4-(prop-1-en-2-yl)benzene
Molecular Weight (g/mol): 152.62
Molecular Formula: C9H9Cl
InChi Key: WQDGTJOEMPEHHL-UHFFFAOYSA-N
SMILES: CC(=C)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results