Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-alpha-methylstyrene (stabilized with TBC) 97.0+%, TCI America™
SDP

Catalog No. C02105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C02105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C02105G Supplier TCI America Supplier No. C02105G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1712-70-5
Molecular Formula C9H9Cl
Molecular Weight (g/mol) 152.621
MDL Number MFCD00000630
InChI Key WQDGTJOEMPEHHL-UHFFFAOYSA-N
Synonym 4-chloro-alpha-methylstyrene, 4-chloroisopropenylbenzene, benzene, 1-chloro-4-1-methylethenyl, 1-chloro-4-prop-1-en-2-yl benzene, 4-isopropenyl-chlorobenzene, p-chloro-alpha-methylstyrene, 4-chloro-a-methylstyrene, 4-isopropenylchlorobenzene, 4-chloro-.alpha.-methylstyrene, acmc-1bnhn
PubChem CID 74381
IUPAC Name 1-chloro-4-prop-1-en-2-ylbenzene
SMILES CC(=C)C1=CC=C(C=C1)Cl

Specifications

Color Yellow
Boiling Point 207°C
Quantity 5 g
UN Number 3082
Formula Weight 152.62
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloro-alpha-methylstyrene (stabilized with TBC)
EINECSNumber (3)-0043&(9)-0452
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.