accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1713-33-3

IUPAC Name: 1-methyl-7-oxabicyclo[4.1.0]heptane
Synonyms: 1-methyl-7-oxabicyclo[4.1.0]heptane
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: FIEKVYPYFQSFTP-UHFFFAOYNA-N
SMILES: CC12CCCCC1O2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results