accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FClCl

CAS RN 1717-00-6

IUPAC Name: 1,1-dichloro-1-fluoroethane
Synonyms: 1,1-dichloro-1-fluoroethane
Molecular Weight (g/mol): 116.94
Molecular Formula: C2H3Cl2F
InChi Key: FRCHKSNAZZFGCA-UHFFFAOYSA-N
SMILES: CC(F)(Cl)Cl
Element 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)