accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1717-59-5

IUPAC Name: 2,2-difluoro-2-(fluorosulfonyl)acetic acid
Synonyms: difluoro(fluorosulfonyl)acetic acid
Molecular Weight (g/mol): 178.08
Molecular Formula: C2HF3O4S
InChi Key: VYDQUABHDFWIIX-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)S(F)(=O)=O
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)