accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17174-98-0

H2NON

CAS RN 17174-98-0

IUPAC Name: 2-cyanobenzamide
Synonyms: benzamide, 2-cyano-
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: STQPCKPKAIRSEL-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC=C1C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results