Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyanobenzamide 96.0+%, TCI America™
SDP

Catalog No. C04315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C04315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C04315G Supplier TCI America Supplier No. C04315G

Chemical Identifiers

CAS 17174-98-0
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.149
MDL Number MFCD00014797
InChI Key STQPCKPKAIRSEL-UHFFFAOYSA-N
Synonym benzamide, 2-cyano, o-cyanobenzamide, benzamide, o-cyano, 4-09-00-03268 beilstein handbook reference, benzamide, o-cyano-8ci, benzamide, 2-cyano-9ci
PubChem CID 72883
IUPAC Name 2-cyanobenzamide
SMILES C1=CC=C(C(=C1)C#N)C(=O)N

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 146.15
Percent Purity ≥96.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyanobenzamide
RTECSNumber CV2483500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.