CAS RN 171744-11-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ GGTI-286
A potent, cell-permeable, and selective inhibitor of GGTase I.