Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ GGTI-286

Catalog No. 345878250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-587-8250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-587-8250UG Supplier MilliporeSigma™ Supplier No. 345878250UG
Add to Cart
Add to Cart

A potent, cell-permeable, and selective inhibitor of GGTase I.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a trifluoroacetate salt.
Description 90% by HPLC
Target Proteins Geranylgeranyltransferase (GGTase)
CAS 171744-11-9
Molecular Formula C23H31N3O3S
Formula Weight 429.6g/mol
Solubility DMSO
Form Glassy Solid
Quantity 250 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.