CAS RN 171778-09-9
CAS RN 171778-09-9
IUPAC Name:
tert-butyl (3S)-3-carbamoyl-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate
Synonyms:
tert-butyl (3S)-3-carbamoyl-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoate
Molecular Weight (g/mol):
410.47
Molecular Formula:
C23H26N2O5
InChi Key:
PBMBMXLGDKBTSS-IBGZPJMESA-N
SMILES:
CC(C)(C)OC(=O)C[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(N)=O
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-Asp OBut -NH2, Advanced ChemTech
CAS: 171778-09-9 Molecular Formula: C23H26N2O5 Molecular Weight (g/mol): 410.47 MDL Number: MFCD00235895 InChI Key: PBMBMXLGDKBTSS-IBGZPJMESA-N IUPAC Name: tert-butyl (3S)-3-carbamoyl-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate SMILES: CC(C)(C)OC(=O)C[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(N)=O