accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1719-53-5

IUPAC Name: dichlorodiethylsilane
Synonyms: dichlorodiethylsilane
Molecular Weight (g/mol): 157.11
Molecular Formula: C4H10Cl2Si
InChi Key: BYLOHCRAPOSXLY-UHFFFAOYSA-N
SMILES: CC[Si](Cl)(Cl)CC
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)