Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Palmitic Acid

Catalog No. 277341KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-734-1KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 27-734-1KG Supplier Sigma Aldrich Supplier No. 277341KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Palmitic Acid

Specifications

CAS 1719-53-5
Density 1.05 g/mL at 25°C
Boiling Point 125°C to 131°C
Molecular Formula C4H10Cl2Si
Linear Formula (C2H5)2SiCl2
MDL Number MFCD00000493
Quantity 1 kg
Synonym 1-Pentadecanecarboxylic acid; C16:0; Cetylic acid; Hexadecanoic acid; NSC 5030; PamOH
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.