accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3COCH3OOOOCH3OCH3OH3C

CAS RN 1719-88-6

IUPAC Name: 3,4,5-trimethoxybenzoyl 3,4,5-trimethoxybenzoate
Synonyms: 3,4,5-trimethoxybenzoyl 3,4,5-trimethoxybenzoate
Molecular Weight (g/mol): 406.39
Molecular Formula: C20H22O9
InChi Key: LQJFTZJNDJDCKV-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1OC)C(=O)OC(=O)C1=CC(OC)=C(OC)C(OC)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results