Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trimethoxybenzoic Anhydride 95.0+%, TCI America™
SDP

Catalog No. T10935G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10935G 5 g
T109325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10935G Supplier TCI America Supplier No. T10935G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1719-88-6
Molecular Formula C20H22O9
Molecular Weight (g/mol) 406.387
MDL Number MFCD00017155
InChI Key LQJFTZJNDJDCKV-UHFFFAOYSA-N
PubChem CID 74398
IUPAC Name (3,4,5-trimethoxybenzoyl) 3,4,5-trimethoxybenzoate
SMILES COC1=CC(=CC(=C1OC)OC)C(=O)OC(=O)C2=CC(=C(C(=C2)OC)OC)OC

Specifications

Melting Point 159°C
Color White
Quantity 5 g
Formula Weight 406.39
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trimethoxybenzoic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.