CAS RN 171905-37-6
CAS RN 171905-37-6
IUPAC Name:
(9H-fluoren-9-yl)methyl (2R)-2-(carbonochloridoyl)pyrrolidine-1-carboxylate
Synonyms:
9H-fluoren-9-ylmethyl (2R)-2-(carbonochloridoyl)pyrrolidine-1-carboxylate
Molecular Weight (g/mol):
355.82
Molecular Formula:
C20H18ClNO3
InChi Key:
DKNLJWSDGPXALN-GOSISDBHSA-N
SMILES:
ClC(=O)[C@H]1CCCN1C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-D-Pro-Cl, Advanced ChemTech
CAS: 171905-37-6 Molecular Formula: C20H18ClNO3 Molecular Weight (g/mol): 355.82 InChI Key: DKNLJWSDGPXALN-GOSISDBHSA-N IUPAC Name: (9H-fluoren-9-yl)methyl (2R)-2-(carbonochloridoyl)pyrrolidine-1-carboxylate SMILES: ClC(=O)[C@H]1CCCN1C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2