CAS RN 171927-40-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Ras/Rac Transformation Blocker, SCH 51344
Ras/Rac Transformation Blocker, SCH 51344, CAS 171927-40-5, is a blocker of Ras-induced malignant transformation and Ras/Rac-dependent dissociation of EMS1 from the actin-myosin II complex.