accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1722-10-7

IUPAC Name: 3-chloro-6-methoxypyridazine
Synonyms: 3-chloro-6-methoxypyridazine
Molecular Weight (g/mol): 144.56
Molecular Formula: C5H5ClN2O
InChi Key: XBJLKXOOHLLTPG-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)N=N1
Quantity 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)