CAS RN 17224-18-9
IUPAC Name:
1-(4-{[(4-butoxyphenyl)methylidene]amino}phenyl)ethan-1-one
Synonyms:
1-(4-{[(4-butoxyphenyl)methylidene]amino}phenyl)ethanone
Molecular Weight (g/mol):
295.38
Molecular Formula:
C19H21NO2
InChi Key:
OCSNPDHAHJRADR-UHFFFAOYSA-N
SMILES:
CCCCOC1=CC=C(C=NC2=CC=C(C=C2)C(C)=O)C=C1
1
–
1
of
1
results
1
N-(4-Butoxybenzylidene)-4-acetylaniline 98.0+%, TCI America™
CAS: 17224-18-9 Molecular Formula: C19H21NO2 Molecular Weight (g/mol): 295.38 MDL Number: MFCD00059418 InChI Key: OCSNPDHAHJRADR-UHFFFAOYSA-N PubChem CID: 533688 IUPAC Name: 1-(4-{[(4-butoxyphenyl)methylidene]amino}phenyl)ethan-1-one SMILES: CCCCOC1=CC=C(C=NC2=CC=C(C=C2)C(C)=O)C=C1