Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Butoxybenzylidene)-4-acetylaniline 98.0+%, TCI America™
SDP

Catalog No. B03721G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B03721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B03721G Supplier TCI America Supplier No. B03721G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17224-18-9
Molecular Formula C19H21NO2
Molecular Weight (g/mol) 295.38
MDL Number MFCD00059418
InChI Key OCSNPDHAHJRADR-UHFFFAOYSA-N
PubChem CID 533688
IUPAC Name 1-(4-{[(4-butoxyphenyl)methylidene]amino}phenyl)ethan-1-one
SMILES CCCCOC1=CC=C(C=NC2=CC=C(C=C2)C(C)=O)C=C1

Specifications

Melting Point 120°C
Color Yellow
Quantity 1 g
Formula Weight 295.38
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Butoxybenzylidene)-4-acetylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.