accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOO

CAS RN 17241-59-7

IUPAC Name: 2-(hydroxymethyl)-9,10-dihydroanthracene-9,10-dione
Synonyms: 2-(hydroxymethyl)anthraquinone
Molecular Weight (g/mol): 238.24
Molecular Formula: C15H10O3
InChi Key: JYKHAJGLEVKEAA-UHFFFAOYSA-N
SMILES: OCC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results