accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOH3COOCH3

CAS RN 1726-23-4

IUPAC Name: 1,2,4-tributyl benzene-1,2,4-tricarboxylate
Synonyms: 1,2,4-tributyl benzene-1,2,4-tricarboxylate
Molecular Weight (g/mol): 378.47
Molecular Formula: C21H30O6
InChi Key: RJIFVNWOLLIBJV-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C1=CC=C(C(=O)OCCCC)C(=C1)C(=O)OCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results