Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyl Trimellitate 98.0+%, TCI America™
SDP

Catalog No. B088025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B088025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B088025ML Supplier TCI America Supplier No. B088025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1726-23-4
Molecular Formula C21H30O6
Molecular Weight (g/mol) 378.47
MDL Number MFCD00059424
InChI Key RJIFVNWOLLIBJV-UHFFFAOYSA-N
Synonym 1,2,4-Benzenetricarboxylic Acid Tributyl Ester, Trimellitic Acid Tributyl Ester
PubChem CID 74411
IUPAC Name 1,2,4-tributyl benzene-1,2,4-tricarboxylate
SMILES CCCCOC(=O)C1=CC=C(C(=O)OCCCC)C(=C1)C(=O)OCCCC

Specifications

Color Yellow
Boiling Point 220°C
Quantity 25 mL
Formula Weight 378.47
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tributyl Trimellitate
EINECSNumber (3)-1372&(3)-3655
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.