accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHHN

CAS RN 172603-05-3

IUPAC Name: tert-butyl N-(piperidin-3-yl)carbamate
Synonyms: tert-butyl N-(piperidin-3-yl)carbamate
Molecular Weight (g/mol): 200.28
Molecular Formula: C10H20N2O2
InChi Key: WUOQXNWMYLFAHT-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC1CCCNC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (2)