accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1728-18-3

CH3OOCH3

CAS RN 1728-18-3

IUPAC Name: 3,3-dimethoxycyclohex-1-ene
Synonyms: 3,3-dimethoxycyclohex-1-ene
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: OVIPECWZMIWWAS-UHFFFAOYSA-N
SMILES: COC1(CCCC=C1)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results