Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyclohexen-1-one Dimethylketal 88.0+%, TCI America™
SDP

Catalog No. C088325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C088325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C088325ML Supplier TCI America Supplier No. C088325ML

Chemical Identifiers

CAS 1728-18-3
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00059564
InChI Key OVIPECWZMIWWAS-UHFFFAOYSA-N
Synonym 3,3-Dimethoxy-1-cyclohexene
PubChem CID 573595
IUPAC Name 3,3-dimethoxycyclohex-1-ene
SMILES COC1(CCCC=C1)OC

Specifications

Color Colorless
Boiling Point 67°C
UN Number 3271
Quantity 25 mL
Formula Weight 142.20
Percent Purity ≥88.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyclohexen-1-one Dimethylketal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.