accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NFBr

CAS RN 17282-01-8

IUPAC Name: 3-bromo-2-fluoro-5-methylpyridine
Synonyms: 3-bromo-2-fluoro-5-methylpyridine
Molecular Weight (g/mol): 190.02
Molecular Formula: C6H5BrFN
InChi Key: FWKQBHMQYXTOTD-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=C(F)N=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results