accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17282-02-9

IUPAC Name: 3-bromo-5-methyl-1,2-dihydropyridin-2-one
Synonyms: 3-bromo-5-methyl-1H-pyridin-2-one
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: KHBRMXVUQOVORD-UHFFFAOYSA-N
SMILES: CC1=CNC(=O)C(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results