Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-methyl-2-pyridone 98.0+%, TCI America™
SDP

Catalog No. B33505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33505G 5 g
B33501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33505G Supplier TCI America Supplier No. B33505G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 17282-02-9 | C6H6BrNO | 188.024 g/mol

Chemical Identifiers

CAS 17282-02-9
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD03791312
InChI Key KHBRMXVUQOVORD-UHFFFAOYSA-N
Synonym 3-bromo-2-hydroxy-5-methylpyridine, 3-bromo-5-methylpyridin-2-ol, 3-bromo-5-methylpyridin-2 1h-one, 3-bromo-2-hydroxy-5-picoline, 3-bromo-5-methyl-pyridin-2-ol, 3-bromo-5-methyl-2 1h-pyridinone, 3-bromo-5-methyl-2-pyridone, 3-bromo-5-methyl-2-pyridinol, 2 1h-pyridinone, 3-bromo-5-methyl, 3-bromo-2-hydroxy-5-picoline 3-bromo-2-hydroxy-5-methylpyridine
PubChem CID 3725261
IUPAC Name 3-bromo-5-methyl-1H-pyridin-2-one
SMILES CC1=CNC(=O)C(=C1)Br

Specifications

Melting Point 161°C
Color White-Yellow
Quantity 5 g
Formula Weight 188.02
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-methyl-2-pyridone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.