accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1729-99-3

IUPAC Name: 8-bromonaphthalene-1-carboxylic acid
Synonyms: 8-bromonaphthalene-1-carboxylic acid
Molecular Weight (g/mol): 251.08
Molecular Formula: C11H7BrO2
InChi Key: DMEZDDHJCUHENA-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C(Br)=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results