Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Bromo-1-naphthoic acid, Technical Grade, Maybridge™

Catalog No. BTB12326EA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB12326EA 10 g Amber Glass Bottle
BTB12326DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB12326EA Supplier Thermo Scientific Chemicals Supplier No. BTB12326EA
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1729-99-3
Molecular Formula C11H7BrO2
Molecular Weight (g/mol) 251.079
InChI Key DMEZDDHJCUHENA-UHFFFAOYSA-N
Synonym 8-bromo-1-naphthoic acid, 1-naphthalenecarboxylic acid, 8-bromo, maybridge1_002138, 8-bromo-1-carboxynaphthalene, 8-bromo-1-naphthalenecarboxylic acid, 1-naphthalenecarboxylicacid, 8-bromo, 8-bromo-naphthalene-1-carboxylic acid
PubChem CID 72873
IUPAC Name 8-bromonaphthalene-1-carboxylic acid
SMILES C1=CC2=C(C(=C1)C(=O)O)C(=CC=C2)Br

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 251.08
Percent Purity 95%
Chemical Name or Material 8-Bromo-1-naphthoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.