accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OON

CAS RN 172998-55-9

IUPAC Name: N-benzyl-2-methoxy-N-phenylbenzamide
Synonyms: N-benzyl-2-methoxy-N-phenylbenzamide
Molecular Weight (g/mol): 317.39
Molecular Formula: C21H19NO2
InChi Key: AAQVKEUFERGCEO-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(=O)N(CC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results