accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1730-25-2

IUPAC Name: bromo(prop-2-en-1-yl)magnesium
Synonyms: allylmagnesium bromide
Molecular Weight (g/mol): 145.28
Molecular Formula: C3H5BrMg
InChi Key: FEMBXICCJNZMMC-UHFFFAOYSA-M
SMILES: Br[Mg]CC=C
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)