Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Allylmagnesium Bromide Solution

Catalog No. 225754800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
800 mL
18 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-575-4800ML 800 mL
22-575-4100ML 100 mL
22-575-418L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 22-575-4800ML Supplier Sigma Aldrich Supplier No. 225754800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Allylmagnesium bromide solution can be used in the Grignard reaction, which is a key step to synthesize:2-(9,10-dihydro-9,10-propanoanthracen-9-yl)-N-methylethanamine (2), a homolog of benzoctamine tarchonanthuslactone(+)-preussinpolyhydroxylated quinolizidine alkaloids

Specifications

CAS 1730-25-2
Density 0.851 g/mL at 25°C
Molecular Formula C3H5BrMg
Linear Formula CH2=CHCH2MgBr
Quantity 800 mL
MDL Number MFCD00000044
Molecular Weight (g/mol) 145.28 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.