accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3

CAS RN 17301-94-9

IUPAC Name: 4-methylnonane
Synonyms: nonane, 4-methyl-
Molecular Weight (g/mol): 142.29
Molecular Formula: C10H22
InChi Key: IALRSQMWHFKJJA-UHFFFAOYNA-N
SMILES: CCCCCC(C)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results