Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylnonane 96.0+%, TCI America™
SDP

Catalog No. M02835ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M02835ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M02835ML Supplier TCI America Supplier No. M02835ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17301-94-9
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00027289
InChI Key IALRSQMWHFKJJA-UHFFFAOYSA-N
Synonym nonane, 4-methyl, 4-n-methylnonane, 4-methylnonane, dl, acmc-209e5n, 4-methylnonane, n-c3h7ch ch3 ch2 4ch3
PubChem CID 28455
IUPAC Name 4-methylnonane
SMILES CCCCCC(C)CCC

Specifications

Color Colorless
Boiling Point 165°C
Quantity 5 mL
UN Number 3295
Formula Weight 142.29
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methylnonane
EINECSNumber (2)-0010
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.